Skip to Content
Merck
All Photos(1)

Documents

Safety Information

Z678449

Krytox® vacuum oil

grade, 1506

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161501
NACRES:
NB.79

mol wt

2160

feature

grade 1506

manufacturer/tradename

Du Pont

viscosity number

(20 °C) (62 mm2/s)

density

1.88 g/mL at 20 °C

Looking for similar products? Visit Product Comparison Guide

General description

DuPont Krytox vacuum pump fluids are used in applications where conventional vacuum pump oils cause safety, waste disposal and maintenance problems. They are nonflammable and eliminate the chance of fire in pumps. They are nonreactive and safe to use in oxygen systems. This gives Krytox superior stability as a vacuum pump fluid.

Krytox vacuum fluids are precisely distilled to provide low vapor pressures and give superior performance.
  • Nonflammable
  • Nonreactive
  • Free from acetal groups
  • Recyclable

Legal Information

Krytox is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

Z678449-BULK:
Z678449-500G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service