Key Documents
Z675075
Stuart heat block for round bottom flasks
robust aluminum block transforms hotplates into heating mantles
About This Item
UNSPSC Code:
41102406
material
aluminum
packaging
pkg of 1 ea
manufacturer/tradename
Stuart SB16/6
General description
This robust aluminum block transforms hotplates into heating mantles. Can be used with any hotplate that has 160 mm plate size. Four different sized holes for 25 mL, 50 mL, 100 mL and 250 mL round bottom flasks. Pre-drilled hole for thermometer or temperature probe.
  • Accommodates 25 mL, 50 mL, 100 mL and 250 mL flasks
  • Fits any 160 mm hotplate
  • Aluminum allows fast consistent heat transfer
  • Four feet secure block to hotplate
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)