Key Documents
Z671983
Hanna portable waterproof dissolved oxygen meter, model HI9142
battery power 4 AA, 500 hr life
About This Item
UNSPSC Code:
41113319
manufacturer/tradename
Hanna
General description
HI 9142 is a rugged, waterproof meter that solves the common problems of field use, such as cold, rain, snow and dust that can damage a meter, rapidly deteriorating its performance and life.
  • Waterproof
  • Simple to calibrate
  • Built-in thermister for temperature management
  • Polarographic probe with 4 m cable
Choose from one of the most recent versions:
Certificates of Analysis (COA)