Z557595
Aldrich® pear-shaped splash-guard adapter
capacity 250 mL, female joint: ST/NS 24/40, male joint: ST/NS 14/20
All Photos(1)
About This Item
Joint
female joint: ST/NS24/40
male joint: ST/NS14/20
description
anti-climb
capacity
250 mL
General description
Application: for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping. Tapered sidewalls eliminate hold up common to round style adapters and permits continuous washing with condensed solvent.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC