Key Documents
Z305502
Sigma-Aldrich
Elite safety Goggles
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
manufacturer/tradename
Bollé
availability
not available in USA and Canada
General description
No Fog PC lens fits over most prescription spectacles with exceptional panoramic vision and comfort to the wearer. This model is fitted with a foam band, but without ventilation holes to give protection against gases and vapours as well as dust. Fits over most prescription spectacles. BS2092.1CDG
Choose from one of the most recent versions:
Certificates of Analysis (COA)