Documents
Z286079
Sigma-Aldrich
Ace vacuum manifold system
#15 Ace-Threds
About This Item
UNSPSC Code:
40161501
NACRES:
NB.79
manufacturer/tradename
Ace Glass
Related Categories
General description
Set includes size-matched components with #15 Ace-Threds for easy assembly of vacuum system. Order 1/2 in. I.D. vacuum tubing separately.
  • 1 Vacuum manifold, dual bank, 3-ports with 5-ring medium hose barb connections, and greaseless, 0 to 4 mm, double action, high-vacuum PTFE stopcocks
  • 1 Mineral oil bubbler, adjustable
  • 1 Vacuum trap, adjustable
  • 4 UHDPE tubing connectors for ½ in. I.D. vacuum tubing
  • 2 Nylon plugs