Key Documents
Z250902
Sigma-Aldrich
InsulEze® insulating kit
Combination
About This Item
UNSPSC Code:
41112303
NACRES:
NB.76
manufacturer/tradename
Glas-Col 100GIK240
General description
Includes one each of kits A, B, and C.
Make insulating less time-consuming, awkward, and unsightly in the lab. Use for heat or cold shielding, insulating over heaters, or as shock absorber. Insulating materials can be cut and shaped to almost any size with minimum fraying of the edges. Material is non-hazardous, non-flammable, and inert to most chemicals.
Legal Information
InsulEze is a registered trademark of Glas-Col Laboratory Products
Choose from one of the most recent versions:
Certificates of Analysis (COA)