WHA5305125
Whatman® Reeve Angel™ filter paper, Grade 305
circles, diam. 125 mm (4.9 in.), pkg of 100 ea
Synonym(s):
Whatman filter, Whatman paper, Z762032, paper filter
About This Item
Recommended Products
material
cellulose
circles
smooth white cellulose
feature
Grade 305
packaging
pkg of 100 ea
manufacturer/tradename
Whatman 5305-125
parameter
fast flow rate
fast flow rate
diam.
125 mm (4.9 in.)
pore size
20-25 μm
basis weight
185
Looking for similar products? Visit Product Comparison Guide
General description
Grade 305 provides good retention, fast filtration speed and has 0.01% typical ash content. A good choice for large particles and gelatinous precipitates.
Note; These papers should not be used for kjeldahl nitrogen analysis.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service