Skip to Content
Merck
All Photos(1)

Documents

W509663

Sigma-Aldrich

Methyl (methylthio)acetate

≥98%

Synonym(s):

Methyl (methylmercapto)acetate, Methyl S-methylthioglycolate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2COOCH3
CAS Number:
Molecular Weight:
120.17
Beilstein:
1902431
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
12.146

Assay

≥98%

refractive index

n20/D 1.4650 (lit.)

bp

49-51 °C/12 mmHg (lit.)

density

1.11 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CSC

InChI

1S/C4H8O2S/c1-6-4(5)3-7-2/h3H2,1-2H3

InChI key

HZYCAKGEXXKCDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service