Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

W396103

Sigma-Aldrich

2-Propylpyrazine

97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
FEMA Number:
3961
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.142
NACRES:
NA.21
Organoleptic:
green; nutty
grade:
FG
Halal
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

grade

FG
Halal

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

97%

refractive index

n20/D 1.494

density

0.975 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; nutty

SMILES string

CCCc1cnccn1

InChI

1S/C7H10N2/c1-2-3-7-6-8-4-5-9-7/h4-6H,2-3H2,1H3

InChI key

DJLLTFRHLPVCEL-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

W396103-BULK:
W396103-VAR:
W396103-100G:
W396103-SAMPLE:
W396103-500G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service