Skip to Content
Merck
All Photos(1)

Key Documents

W384606

Sigma-Aldrich

Litsea cubeba oil

Certified organic (NOP)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3846
MDL number:
UNSPSC Code:
12164502
Organoleptic:
grassy; lemon
grade:
Certified organic (NOP)
Halal
Kosher
food allergen:
no known allergens

grade

Certified organic (NOP)
Halal
Kosher

Quality Level

origin

Vietnam origin

refractive index

n20/D 1.480 (lit.)

bp

232 °C/760 mmHg (lit.)

density

0.884 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

grassy; lemon

Looking for similar products? Visit Product Comparison Guide

Legal Information

Certified organic by MOSA

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

130.0 °F - closed cup

Flash Point(C)

54.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service