Skip to Content
Merck
All Photos(1)

Documents

Safety Information

W324801

Sigma-Aldrich

2-Vinylanisole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCH3)CHCH2
CAS Number:
Molecular Weight:
134.18
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

Assay

98%

contains

(may contain up to 0.10% t-butylcatechol, synthetic as stabilzer)

refractive index

n20/D 1.5540 (lit.)

bp

36-43 °C/0.5 mmHg (lit.)

density

0.999 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

green

SMILES string

COc1ccccc1C=C

InChI

1S/C9H10O/c1-3-8-6-4-5-7-9(8)10-2/h3-7H,1H2,2H3

InChI key

SFBTTWXNCQVIEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

135.0 °F - closed cup

Flash Point(C)

57.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

W324801-VAR:
W324801-BULK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service