W294926
Propyl hexanoate
natural, ≥95%, FG
Synonym(s):
n-Propyl hexanoate, Caproic acid propyl ester, Propyl caproate
About This Item
Recommended Products
grade
FG
Fragrance grade
Halal
Kosher
natural
Agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
Assay
≥95%
refractive index
n20/D 1.412 (lit.)
bp
187 °C (lit.)
mp
−69 °C (lit.)
density
0.867 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
green; tropical; fruity; pineapple; sweet
SMILES string
CCCCCC(=O)OCCC
InChI
1S/C9H18O2/c1-3-5-6-7-9(10)11-8-4-2/h3-8H2,1-2H3
InChI key
HTUIWRWYYVBCFT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
125.1 °F - closed cup
Flash Point(C)
51.7 °C - closed cup
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
FSL
Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid
JAN Code
W294926-1KG-K:4548173974446
W294926-VAR-K:
W294926-100G:
W294926-1KG:
W294926-SAMPLE:
W294926-BULK-K:
W294926-100G-K:4548173974439
W294926-25G:
W294926-SAMPLE-K:
W294926-25G-K:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service