Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

W282928

Sigma-Aldrich

Orris concrete

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2829
MDL number:
UNSPSC Code:
12164502

biological source

Iris florentina, Iris germanica
synthetic

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117

composition

iron, 15%

origin

France origin

mp

40-46 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apricot; berry; grape; honey; oily; woody; peach; raspberry; sweet; vanilla; violet

Biochem/physiol Actions

Taste at 5 ppm in 5%

Preparation Note

Extraction method: steam distillation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

W282928-BULK:
W282928-100G:
W282928-SAMPLE-K:
W282928-25G-K:
W282928-25G:
W282928-100G-K:
W282928-BULK-K:
W282928-SAMPLE:
W282928-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 24, 27-27 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service