Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

W280712

Sigma-Aldrich

Octyl butyrate

natural (US), ≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2(CH2)7CH3
CAS Number:
Molecular Weight:
200.32
FEMA Number:
2807
EC Number:
Council of Europe no.:
272c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.046

grade

Kosher
natural (US)

Assay

≥95%

refractive index

n20/D 1.425 (lit.)

bp

224 °C (lit.)

mp

−56 °C (lit.)

density

0.862 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; musty; orange; citrus; fruity

SMILES string

CCCCCCCCOC(=O)CCC

InChI

1S/C12H24O2/c1-3-5-6-7-8-9-11-14-12(13)10-4-2/h3-11H2,1-2H3

InChI key

PWLNAUNEAKQYLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

W280712-BULK-K:
W280712-SAMPLE-K:
W280712-100G:
W280712-25G-K:
W280712-100G-K:
W280712-1KG-K:
W280712-25G:
W280712-VAR-K:
W280712-1KG:
W280712-SAMPLE:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service