W206202
α-Amylcinnamaldehyde dimethyl acetal
≥97%, mixture of isomers
Synonym(s):
α-Pentylcinnamaldehyde dimethyl acetal, [2-(Dimethoxymethyl)-1-hepten-1-yl]benzene
About This Item
Assay
≥97%
refractive index
n20/D 1.508 (lit.)
bp
300 °C (lit.)
density
0.959 g/mL at 25 °C (lit.)
SMILES string
CCCCC\C(=C/c1ccccc1)C(OC)OC
InChI
1S/C16H24O2/c1-4-5-7-12-15(16(17-2)18-3)13-14-10-8-6-9-11-14/h6,8-11,13,16H,4-5,7,12H2,1-3H3/b15-13+
InChI key
QCHZKUPVENJLAW-FYWRMAATSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
FSL
Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid
JAN Code
W206202-SAMPLE:
W206202-VAR:
W206202-10KG:
W206202-BULK:
W206202-5KG:
W206202-1KG:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service