W200301
Acetaldehyde
≥99%, meets FCC analytical specification
Synonym(s):
Ethanal
About This Item
Recommended Products
biological source
synthetic
Quality Level
grade
Kosher
reg. compliance
meets FCC analytical specification
vapor density
1.52 (vs air)
vapor pressure
14.63 psi ( 20 °C)
Assay
≥99%
autoignition temp.
365 °F
expl. lim.
60 %
refractive index
n20/D 1.332 (lit.)
bp
21 °C (lit.)
mp
−125 °C (lit.)
density
0.785 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
pungent; ethereal; fruity
storage temp.
2-8°C
SMILES string
CC=O
InChI
1S/C2H4O/c1-2-3/h2H,1H3
InChI key
IKHGUXGNUITLKF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-38.0 °F - closed cup
Flash Point(C)
-38.89 °C - closed cup
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PRTR
Specified Class I Designated Chemical Substances
FSL
Group 4: Flammable liquids
Special flammables
Hazardous rank I
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
W200301-2.4KG:
W200301-2.1KG:
W200301-VAR:
W200301-19.5KG-K:
W200301-PZ:
W200301-12KG:
W200301-SAMPLE-K:
W200301-1.9KG-K:4548173955452
W200301-2.5L:
W200301-16.5KG-K:4548173955490
W200301-2KG-K:4548173955476
W200301-16.5KG:
W200301-BULK-K:
W200301-VAR-K:
W200301-4X2KG:
W200301-100ML-K:
W200301-30KG:
W200301-78G-K:4548173955506
W200301-30KG-K:4548173955483
W200301-SAMPLE:
W200301-2KG:
W200301-1.96KG:
W200301-12KG-K:4548173955469
W200301-BULK:
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service