Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

T43702

Sigma-Aldrich

Triacetin

99%

Synonym(s):

1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glyceryl triacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COOCH2)2CHOCOCH3
CAS Number:
Molecular Weight:
218.20
Beilstein:
1792353
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

7.52 (vs air)

Assay

99%

autoignition temp.

809 °F

expl. lim.

1.05 %, 189 °F
7.73 %, 215 °F

refractive index

n25/D 1.429-1.431 (lit.)

bp

258 °C (lit.)

mp

3 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCC(COC(C)=O)OC(C)=O

InChI

1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3

InChI key

URAYPUMNDPQOKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 3 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

T43702-BULK:
T43702-1L:
T43702-3KG:
T43702-250ML:
T43702-1KG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service