Documents
SYNF644125
Synthware pressure equalizing funnel with high vacuum valve
125 mL, joint: ST/NS 24/40, valve size 0-4 mm
Synonym(s):
Synthware F644125, Addition funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F644125
capacity
125 mL
125 mL
Related Categories
General description
With PTFE extended tip valve for semi-needle valve adjustment.
Legal Information
Synthware is a trademark of Kemtech America Inc.