SLW1470/01M
Pyrex® Crystallizing dish, flat bottom
60 mL
Synonym(s):
Pyrex, SciLabware 1470/01M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
flat bottom
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1470/01M
diam. × H
58 mm × 35 mm
capacity
60 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Complies to DIN 12337
- Manufactured from Pyrex borosilicate glass
- Suitable for use up to 400°C
- Chemically and thermally resistant
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service