Skip to Content
Merck
All Photos(1)

Documents

Safety Information

QBD11471

Sigma-Aldrich

MAL-dPEG®12-Tris(m-dPEG®24)3

>95% (HPLC)

Synonym(s):

Branched Maleimide-PEG-m, Branched mPEG-maleimide, MAL-NH-PEG600-Tris(mPEG1000)3, MAL-PEG12-Tris(m-PEG24)3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C194H378N6O94
Molecular Weight:
4299.06
UNSPSC Code:
12352106
NACRES:
NA.22

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

MAL-dPEG12-Tris(m-dPEG24)3 is a thiol-reactive, methyl-terminated, four-arm monodispersed PEG product designed to modulate biodistribution of conjugates. Tris forms the core of the molecule. Three equal-length methyl-terminated long arms plus one maleimido-terminated short arm extend from the tris core. From the maleimido olefin to the terminal methyl groups on each arm, the end-to-end length of this product is 127 atoms with an average distance of 103.7 Å. The amphiphilic PEG imparts hydrophilicity to conjugates, reduces or eliminates the conjugates′ antigenicity, and increases the hydrodynamic volume of conjugated molecules. Applications for this product include eradicating renal clearance, maintaining in vivo circulation for more extended periods, and suppressing immune responses to conjugates.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

QBD11471-25MG:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiming Ding et al.
Bioconjugate chemistry, 24(11), 1945-1954 (2013-11-02)
The tumor-associated glycoprotein-72 (TAG-72) antigen is highly overexpressed in various human adenocarcinomas and anti-TAG-72 monoclonal antibodies, and fragments are therefore useful as pharmaceutical targeting vectors. In this study, we investigated the effects of site-specific PEGylation with MW 2-4 kDa discrete

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service