L880000-HH
DCI Activator
configured for PerkinElmer 8900, configured for Polygen, 0.25M
Synonym(s):
DCI activator solution
About This Item
Recommended Products
Quality Level
type
for DNA synthesis
product line
Proligo Reagents
packaging
pkg of 6 × 120 mL (L880012-06)
pkg of 6 × 200 mL (L880020-06)
concentration
29-33 g/L (Dicyanoimidazole, titration)
impurities
≤30 ppm water content (Karl Fischer)
color
clear
compatibility
configured for PerkinElmer 8900
configured for Polygen
Looking for similar products? Visit Product Comparison Guide
Preparation Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
41.0 °F
Flash Point(C)
5 °C
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
PDSCL
Deleterious substance
FSL
Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid
ISHL Indicated Name
Substances Subject to be Indicated Names
ISHL Notified Names
Substances Subject to be Notified Names
JAN Code
L032080-200ML:
L880012-06:4548173279404
L032080-6X200ML:4548174010181
L880020-C02-01:
L880020-06:4548173225753
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service