Skip to Content
Merck
All Photos(1)

Documents

Safety Information

J100042

Sigma-Aldrich

Bis(cyclopentadienyl)magnesium(II)

≥99.99% trace metals basis (excludes ~300ppm Al)

Synonym(s):

MgCp2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(C5H5)2
CAS Number:
Molecular Weight:
154.49
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.99% trace metals basis (excludes ~300ppm Al)

form

solid

mp

180 °C (dec.) (lit.)

SMILES string

[Mg].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.Mg/c2*1-2-4-5-3-1;/h2*1-5H;

InChI key

USZGMDQWECZTIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atomic number of base material: 12 Magnesium

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Sol. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 3: Spontaneously combustible substances and water- reactive materials
Organic metal compounds (except alkyl aluminum and alkyl lithium)
Hazardous rank I
1st spontaneously combustible materials and water reactive materials

JAN Code

J100042-BULK:
J100042-10G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service