GF09152052
Indium
foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Synonym(s):
Indium, IN000270
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
vapor pressure
<0.01 mmHg ( 25 °C)
Assay
100%
form
foil
manufacturer/tradename
Goodfellow 091-520-52
resistivity
8.37 μΩ-cm
mp
156.6 °C (lit.)
density
7.3 g/mL at 25 °C (lit.)
SMILES string
[In]
InChI
1S/In
InChI key
APFVFJFRJDLVQX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of chemical physics, 134(10), 104303-104303 (2011-03-17)
The doubly positively charged gas-phase molecules BrO(2+) and NBr(2+) have been produced by prolonged high-current energetic oxygen (17 keV (16)O(-)) ion surface bombardment (ion beam sputtering) of rubidium bromide (RbBr) and of ammonium bromide (NH(4)Br) powdered ionic salt samples, respectively
The Analyst, 138(16), 4463-4469 (2013-04-05)
In order to fully characterize the zinc white artists' pigment (ZnO), much used since the mid-nineteenth century, three samples collected in the early 20th century were studied using a combination of synchrotron and macroscopic photoluminescence spectroscopy and imaging. An improved
Radiation protection dosimetry, 144(1-4), 575-579 (2010-11-11)
At the Nuclear Fuel Cycle Engineering Laboratories of the Japan Atomic Energy Agency (JAEA), small pieces of indium foil incorporated into personal dosemeters have been used for personnel screening in criticality accidents. Irradiation tests of the badges were performed using
Evaluation of the repair of mechanical furcation perforations using amalgam, gutta-percha, or indium foil.
Journal of endodontics, 12(6), 249-256 (1986-06-01)
Anatomic repair of internal perforations with indium foil and silver amalgam: outline of a method.
The New York journal of dentistry, 39(10), 454-457 (1969-12-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service