DWK211184806
DURAN® Super duty high form beaker with spout
glass, capacity (600 mL)
Synonym(s):
211184806
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
glass
packaging
case of 10 ea
manufacturer/tradename
DURAN® 211184806
H × diam.
150 cm × 80 cm
capacity
(600 mL)
Looking for similar products? Visit Product Comparison Guide
General description
Higher impact strength, mechanical strength and stability achieved by reinforced rim. Economical due to longer life span and outstanding corrosion protection. Spout for clean pouring. White easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white ceramic print.
Other Notes
Browse all glass beakers
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service