Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

D89204

Sigma-Aldrich

3-(Diethylamino)propylamine

≥99%

Synonym(s):

N,N-Diethyl-1,3-diaminopropane, N,N-Diethyl-1,3-propanediamine, DEAPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2N(CH2)3NH2
CAS Number:
Molecular Weight:
130.23
Beilstein:
741879
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.4 (vs air)

vapor pressure

19.5 mmHg ( 70 °C)

Assay

≥99%

form

liquid

autoignition temp.

419 °F

expl. lim.

11.6 %

refractive index

n20/D 1.4416 (lit.)

bp

159 °C (lit.)

density

0.826 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CCCN

InChI

1S/C7H18N2/c1-3-9(4-2)7-5-6-8/h3-8H2,1-2H3

InChI key

QOHMWDJIBGVPIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Diethylamino)propylamine (3-(diethylamino)-1-propylamine) is a diamine.

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water soluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

D89204-500ML:
D89204-25ML:
D89204-1L:
D89204-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical surface modifications of microfibrillated cellulose.
Stenstad P, et al.
Cellulose, 15(1) (2008)
Amine-modified poly (vinyl alcohol) s as non-viral vectors for siRNA delivery: effects of the degree of amine substitution on physicochemical properties and knockdown efficiency.
Nguyen J, et al.
Pharmaceut. Res., 27(12), 2670-2682 (2010)
Hyaluronated nanoparticles with pH-and enzyme-responsive drug release properties.
Kim SW, et al.
Colloids and Surfaces. B, Biointerfaces, 116, 359-364 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service