Skip to Content
Merck
All Photos(1)

Documents

Safety Information

D81009

Sigma-Aldrich

Dicyclopentadiene dioxide, mixture of endo and exo isomers

97%

Synonym(s):

Dicyclopentadiene diepoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

97%

mp

185-189 °C (lit.)

SMILES string

C1C2OC2C3C1[C@H]4C[C@@H]3C5OC45

InChI

1S/C10H12O2/c1-4-3-2-6-10(11-6)7(3)5(1)9-8(4)12-9/h3-10H,1-2H2/t3?,4-,5+,6?,7?,8?,9?,10?/m1/s1

InChI key

BQQUFAMSJAKLNB-CNHCLJDLSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

D81009-VAR:
D81009-BULK:
D81009-5G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service