Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

D197157

Sigma-Aldrich

1-Iodo-2,4-dinitrobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3(NO2)2
CAS Number:
Molecular Weight:
294.00
Beilstein:
1113584
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

86-88 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(I)c(c1)[N+]([O-])=O

InChI

1S/C6H3IN2O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H

InChI key

FXMKXMJLXRTQSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 5: Self-reactive substances
Nitro compounds
Hazardous rank I
1st self-reactive materials

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

D197157-5G:
D197157-VAR:
D197157-1G:
D197157-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Gupta et al.
Mutation research, 381(1), 41-47 (1997-12-24)
The 1-halogen substituted 2,4-dinitrobenzenes have been found to be mutagenic in Salmonella TA98 with an activity order of 1-fluoro > 1-chloro > 1-bromo > 1-iodo. This specific activity was not lowered in the nitroreductase deficient strain TA98NR and O-acetyltransferase deficient
Enamul Hoque et al.
Applied and environmental microbiology, 73(8), 2697-2707 (2007-02-13)
The occurrence and activation of glutathione S-transferase (GST) and the GST activities in biofilms in cold sulfidic spring waters were compared to the occurrence and activation of GST and the GST activities of the aquatic fungal strains EH5 and EH7

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service