Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

CDS003383

Sigma-Aldrich

(5-Methyl-2-furyl)methanol

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

SMILES string

Cc1ccc(CO)o1

InChI

1S/C6H8O2/c1-5-2-3-6(4-7)8-5/h2-3,7H,4H2,1H3

InChI key

VOZFDEJGHQWZHU-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

CDS003383-500MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B M Wherry et al.
Journal of dairy science, 102(5), 3868-3878 (2019-03-05)
Maillard reactions occur in dairy products during heat treatment. Furfuryl alcohol (FA) may be found in dairy products as a result of Maillard reactions. The recent posting in California Proposition 65 indicates that FA may be carcinogenic, and for this
Dandan Pu et al.
Food chemistry, 298, 125003-125003 (2019-07-02)
The oral breakdown, sensory properties, and volatile release during mastication of white bread were investigated. The results of correlation analysis for white bread's physical properties and it's oral physiological parameters during chewing have elucidated that bread's physical properties determined the
Jie Liu et al.
Food chemistry, 345, 128826-128826 (2021-02-20)
Brown sugar (non-centrifugal cane sugar) is popular for its pleasant caramel-like aroma and sweetness. Vacuum simultaneous steam distillation and extraction (V-SDE) and gas chromatography-mass spectrometry (GC-MS) was used to study the volatile fraction of brown sugar. To further determine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service