BMSR2020150
Benchmark RotoMini Rotator Tube holder
to hold, 4 x 1015 mL
Synonym(s):
tube rotator, mini tube rotator, lab roller, mini lab roller, lab revolver, rotoflex, rotoflex, argos rotator, mini rotator
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
to hold 4 x 1015 mL
manufacturer/tradename
Benchmark Scientific R2020-150
Looking for similar products? Visit Product Comparison Guide
Application
Mixing of samples in tubes
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service