Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

97071

Sigma-Aldrich

4-(4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecyloxy)benzyl alcohol

≥97.0%

Synonym(s):

4-(1H,1H,2H,2H,3H,3H-Perfluoroundecyloxy)benzyl alcohol, 4-[3-(Perfluorooctyl)-1-propyloxy]benzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13F17O2
CAS Number:
Molecular Weight:
584.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0%

mp

85-89 °C

SMILES string

OCc1ccc(OCCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc1

InChI

1S/C18H13F17O2/c19-11(20,6-1-7-37-10-4-2-9(8-36)3-5-10)12(21,22)13(23,24)14(25,26)15(27,28)16(29,30)17(31,32)18(33,34)35/h2-5,36H,1,6-8H2

InChI key

PCXONXLXJULKDJ-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

97071-1G-F:
97071-VAR-F:
97071-5G-F:
97071-BULK-F:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service