Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

96018

Sigma-Aldrich

Z-Tyr(Bzl)-OH

≥98.0% (TLC)

Synonym(s):

Z-O-benzyl-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23NO5
CAS Number:
Molecular Weight:
405.44
Beilstein:
3178475
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (TLC)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@H](Cc1ccc(OCc2ccccc2)cc1)NC(=O)OCc3ccccc3

InChI

1S/C24H23NO5/c26-23(27)22(25-24(28)30-17-20-9-5-2-6-10-20)15-18-11-13-21(14-12-18)29-16-19-7-3-1-4-8-19/h1-14,22H,15-17H2,(H,25,28)(H,26,27)/t22-/m0/s1

InChI key

IPAODWFPTVIUSZ-QFIPXVFZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

96018-BULK:
96018-VAR:
96018-5G:
96018-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chloé Guilbaud-Chéreau et al.
ACS applied materials & interfaces, 11(14), 13147-13157 (2019-03-14)
Molecular gels formed by the self-assembly of low-molecular-weight gelators have received increasing interest because of their potential applications in drug delivery. In particular, the ability of peptides and amino acids to spontaneously self-assemble into three-dimensional fibrous network has been exploited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service