Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

95998

Sigma-Aldrich

Z-Asp-OMe

≥98.0% (TLC)

Synonym(s):

N-Cbz-L-aspartic acid α-methyl ester, Z-L-Aspartic acid 1-methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(NHCO2CH2C6H5)CO2CH3
CAS Number:
Molecular Weight:
281.26
Beilstein:
4813538
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

COC(=O)[C@H](CC(O)=O)NC(=O)OCc1ccccc1

InChI

1S/C13H15NO6/c1-19-12(17)10(7-11(15)16)14-13(18)20-8-9-5-3-2-4-6-9/h2-6,10H,7-8H2,1H3,(H,14,18)(H,15,16)/t10-/m0/s1

InChI key

MFFFBNAPQRDRQW-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

95998-VAR:
95998-5G:
95998-BULK:
95998-1G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service