94814
3-(3-Pyridyl)-L-alanine
≥98.0% (HPLC)
Synonym(s):
(S)-2-Amino-3-(3-pyridyl)propionic acid, 3′-Aza-L-phenylalanine
About This Item
Recommended Products
Assay
≥98.0% (HPLC)
form
powder
reaction suitability
reaction type: solution phase peptide synthesis
application(s)
peptide synthesis
SMILES string
N[C@@H](Cc1cccnc1)C(O)=O
InChI
1S/C8H10N2O2/c9-7(8(11)12)4-6-2-1-3-10-5-6/h1-3,5,7H,4,9H2,(H,11,12)/t7-/m0/s1
InChI key
DFZVZEMNPGABKO-ZETCQYMHSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
94814-1G-F:
94814-VAR-F:
94814-BULK-F:
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service