Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

93786

Sigma-Aldrich

(R)-2-Aminooctane

≥98.0% (GC)

Synonym(s):

(R)-2-Octylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(NH2)CH3
CAS Number:
Molecular Weight:
129.24
Beilstein:
1719320
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥98.0% (GC)

form

liquid

optical purity

enantiomeric ratio: >99.0:1.0 (GC)

SMILES string

CCCCCC[C@@H](C)N

InChI

1S/C8H19N/c1-3-4-5-6-7-8(2)9/h8H,3-7,9H2,1-2H3/t8-/m1/s1

InChI key

HBXNJMZWGSCKPW-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F

Flash Point(C)

51 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

JAN Code

93786-1G-F:
93786-VAR-F:
93786-250MG-F:
93786-BULK-F:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service