933449
AFM Disc
disc diam. 15 mm
Synonym(s):
Metal Discs
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
disc diam.
15 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Each metal disc pack contains 50 units of metal discs.
Application
These are high quality metal discs with smooth edges and flat surfaces for use in atomic force microscopy. Diameter: 15mm.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service