Skip to Content
Merck
All Photos(2)

Documents

Safety Information

925209

Sigma-Aldrich

Chiralyst Ru637

Umicore

Synonym(s):

RuCl(p-cymene)[(S,S)-Ts-DPEN], Ru[(S,S)-TsDPEN](p-cymene)Cl, [((S,S)-2-Amino-1,2-diphenylethyl)[(4-tolyl)sulfonyl]amido](chloro)(p-cymene)ruthenium(II), [N-[(1S,2S)-2-(Amino-κN)-1,2-diphenylethyl]-4-methylbenzenesulfonamidato-κN]chloro[(1,2,3,4,5,6-η)-1-methyl-4-(1-methylethyl)benzene]-ruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H35ClN2O2RuS
CAS Number:
Molecular Weight:
636.21
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Quality Level

form

powder

mp

>175 °C

storage temp.

2-8°C

SMILES string

CC(C)c1ccc(C)cc1.Cc2ccc(cc2)S(=O)(=O)N([Ru]Cl)[C@H]([C@@H](N)c3ccccc3)c4ccccc4

InChI

1S/C21H21N2O2S.C10H14.ClH.Ru/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17;1-8(2)10-6-4-9(3)5-7-10;;/h2-15,20-21H,22H2,1H3;4-8H,1-3H3;1H;/q-1;;;+2/p-1/t20-,21-;;;/m0.../s1

InChI key

AZFNGPAYDKGCRB-XCPIVNJJSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst involved in:
  • Asymmetric transfer hydrogenation of imines and ketones
  • Intramolecular asymmetric reductive amination
  • Tandem hydroformylation / hydrogenation of terminal olefins

Reactant involved in studies of thermal decomposition of areneruthenium chiral amido-amine alkyl complexes
RuCl(p-cymene)[(S,S)-Ts-DPEN] is a chiral diamine ligand complexed with ruthenium, which can be used for the asymmetric transfer hydrogenation of a variety of imines.

Storage and Stability

Do not store above 25 °C (77 °F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

Additional information available at www.pmc.umicore.com

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

925209-250MG:
925209-VAR:
925209-BULK:
925209-1G:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service