Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

922463

Sigma-Aldrich

Rhodamine B-PEG2k-N3

average Mn 2,000

Synonym(s):

Fluorescent PEG, RB-PEG, Rhodamine B PEG

Sign Into View Organizational & Contract Pricing


About This Item

Assay

≥90% (HPLC)

form

solid

mol wt

average Mn 2,000 (by NMR)
average Mn 2,000

color

red to purple

Ω-end

azide

α-end

rhodamine B

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Application

Polyethylene glycols (PEGs) are FDA approved hydrophilic polymers. They are non-toxic, non-immunogenic and non-antigenic, and are widely used in a variety of biomedical applications such as bioconjugation, drug delivery, surface functionalization, and tissue engineering. Rhodamine B-PEG2k-N3 is a heterobifunctional PEG with a Rhodamine B fluorescein dye and an azide endgroup. Rhodamine B has peak fluorescence emission at 572-575 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

922463-BULK:
922463-50MG:
922463-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service