917516
Empty cartridges
3 mL, opaque black, 50/box
Synonym(s):
3d bioprinting consumables, Syringe barrels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
description
3mL cartridges (50/box), opaque black, for complete light blockage
diam. × L
11.1 mm (0.44′′) × 73.0 mm (2.88′′)
Application
Empty cartridges for 3D Bioprinting application. The precision fit between the barrel and piston ensures consistent fluid deposition. The wiper piston improves fluid control and prevents fluid from dripping. Pistons, end and tip caps are sold separately.
Opaque black cartridges for complete light blockage
Opaque black cartridges for complete light blockage
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service