Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

91652

Sigma-Aldrich

Triethylphosphine solution

technical, ~70% in isopropanol

Synonym(s):

PEt3, Triethylphosphorous solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3P
CAS Number:
Molecular Weight:
118.16
Beilstein:
969170
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

grade

technical

concentration

~70% in isopropanol

refractive index

n20/D 1.430

SMILES string

CCP(CC)CC

InChI

1S/C6H15P/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

RXJKFRMDXUJTEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Regioselective formation of alcohols from hydrocarbonylation of alkenes
  • Bronsted acid-assisted synthesis of dihydropyrones from aromatic aldehydes and allenoates
  • Silaboration of enynes
  • Conversion of phenylacetic acids to phenylacetonitriles of benzonitriles
  • Thermal decomposition of platinum and palladium naphthalenediyl
  • Asymmetrical Morita-Baylis-Hillman reaction

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

91652-100ML:4548173183275
91652-BULK:
91652-25ML:4548173183282


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service