Skip to Content
Merck
All Photos(1)

Documents

Safety Information

910228

Sigma-Aldrich

1,3,5,7-Tetramethyl-2,4,8-trioxo-6-(2-bromophenyl)-6-phosphaadamantane

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20BrO3P
CAS Number:
Molecular Weight:
371.21
UNSPSC Code:
12352200

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: ligand

mp

151 °C

functional group

phosphine

Application

1,3,5,7-Tetramethyl-2,4,8-trioxo-6-(2-bromophenyl)-6-phosphaadamantane is a precursor to PAd-Dalphos (900593) and related phosphine ligands developed by the Stradiotto lab.

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

910228-BULK:
910228-250MG:
910228-1G:
910228-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher M Lavoie et al.
Nature communications, 7, 11073-11073 (2016-03-24)
Palladium-catalysed C(sp(2))-N cross-coupling (that is, Buchwald-Hartwig amination) is employed widely in synthetic chemistry, including in the pharmaceutical industry, for the synthesis of (hetero)aniline derivatives. However, the cost and relative scarcity of palladium provides motivation for the development of alternative, more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service