Skip to Content
Merck
All Photos(2)

Documents

Safety Information

902918

Sigma-Aldrich

Diisopropyl 2-((tosyloxy)imino)malonate

≥95%

Synonym(s):

O-Tosylated ketomalonate oxime, Kurti doubly electrophilic N-linchpin reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO7S
CAS Number:
Molecular Weight:
371.41
UNSPSC Code:
12352101

Assay

≥95%

form

powder or crystals

mp

82-87 °C

Application

As demonstrated by Laszlo Kürti′s lab, ketomalonate oxime O-sulfonates act as doubly N-electrophilic linchpin reagents towards strong C-nucleophiles such as alkyl- and arylmetals (e.g., Grignard reagents). Thus, symmetrical dialkyl- as well as diarylamines may be directly prepared at low tempretatures and in the absence of transition metal catalysts. Other ketomalonate oxime O-sulfonates include 902705, 902624, and 902748

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

902918-1G:
902918-BULK:
902918-VAR:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Padmanabha V Kattamuri et al.
Journal of the American Chemical Society, 139(32), 11184-11196 (2017-06-27)
Given the importance of amines in a large number of biologically active natural products, active pharmaceutical ingredients, agrochemicals, and functional materials, the development of efficient C-N bond-forming methods with wide substrate scope continues to be at the frontier of research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service