Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

901460

Sigma-Aldrich

Dimethyl (terephthalate-13C8)

≥99 atom % 13C, ≥99% (CP)

Synonym(s):

Dimethyl terephthalate-1,2,3,4,5,6,α,α′-13C8

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C8C2H10O4
CAS Number:
Molecular Weight:
202.13
UNSPSC Code:
12352005

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

form

flakes

mass shift

M+8

SMILES string

O=[13C](OC)[13C]1=[13CH][13CH]=[13C]([13C](OC)=O)[13CH]=[13CH]1

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

901460-BULK:
901460-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service