Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

900972

Sigma-Aldrich

2-(3-Bromo-4-chloro-6-methyl-1H-pyrrolo[3,2-c]pyridin-1-yl)ethan-1-amine dihydrochloride

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11BrClN3 · 2HCl
Molecular Weight:
361.49
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

storage temp.

2-8°C

Application

Pyrrolo[3,2-c] pyridine derivative (5-azaindole) that can be used as an indole bioisostere for designing azaindole-based drugs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

900972-VAR:
900972-BULK:
900972-100MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of tetrahydro-5-azaindoles and 5-azaindoles using Pictet?Spengler reaction?appreciable difference in products using different acid catalysts.
Shumaila A MA, et al
Tetrahedron, 67(5), 936-942 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service