Skip to Content
Merck
All Photos(3)

Key Documents

Safety Information

900348

Sigma-Aldrich

Dicyclohexylamine borane

95%

Synonym(s):

DCAB, DICAB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H26BN
CAS Number:
Molecular Weight:
195.15
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

crystals

reaction suitability

reagent type: reductant

mp

118.2 °C

storage temp.

2-8°C

SMILES string

[H][N+](C1CCCCC1)([BH3-])C2CCCCC2

InChI

1S/C12H26BN/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12,14H,1-10H2,13H3

InChI key

FWNIZOBKEDZMFC-UHFFFAOYSA-N

Application

Dicyclohexylamine borane complex applied by the Pucheault Group at the Universite de Bordeaux as a cost effective and easy to handle alternative to current borylation reagents. In one pot, aryl and vinyl halides can be converted to the boronic acid derivative of your choice (BF3K, Bpin, MIDA, and others) depending on quench and workup.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

900348-VAR:
900348-1G:
900348-BULK:
900348-5G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service