Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

86055

Sigma-Aldrich

4-Sulfamoylbutyric acid

technical, ≥90% (T)

Synonym(s):

3-Carboxypropanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2S(=O)2CH2CH2CH2COOH
CAS Number:
Molecular Weight:
167.18
Beilstein:
8254788
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (T)

form

crystals

reaction suitability

reagent type: cross-linking reagent

impurities

≤5% water

mp

~90 °C (dec.)

functional group

carboxylic acid
sulfonamide

SMILES string

O=S(N)(CCCC(O)=O)=O

InChI

1S/C4H9NO4S/c5-10(8,9)3-1-2-4(6)7/h1-3H2,(H,6,7)(H2,5,8,9)

InChI key

OMPGKWICGUBROA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

86055-500MG:
86055-VAR:
86055-2.5G:
86055-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service