82817
Pyridinium p-toluenesulfonate polymer-bound
extent of labeling: ~3.5 mmol/g toluene 4-sulfonate loading
Synonym(s):
p-Toluenesulfonic acid pyridine salt polymer-bound, Poly(4-vinylpyridinium (p-toluenesulfonate)) crosslinked with 2% divinylbenzene divinylbenzene
About This Item
Recommended Products
form
lumps
extent of labeling
~3.5 mmol/g toluene 4-sulfonate loading
matrix
crosslinked with 2% DVB
particle size
100-200 mesh
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
JAN Code
82817-10G:
82817-INTR:
82817-VAR:
82817-BULK:
82817-50G:
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service