Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

804452

Sigma-Aldrich

Potassium 3-ethoxycarbonylbenzoyltrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9BF3KO3
CAS Number:
Molecular Weight:
284.08
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

powder

SMILES string

CCOC(C1=CC=CC(C(B(F)(F)F)=O)=C1)=O.[K]

InChI

1S/C10H9BF3O3.K.H/c1-2-17-10(16)8-5-3-4-7(6-8)9(15)11(12,13)14;;/h3-6H,2H2,1H3;;

InChI key

WJLIBYVPNAKYHP-UHFFFAOYSA-N

Application

Acyltrifluroborates (KAT′s) are bench, air, and moisture stable reagents for rapid, chemoselective amide bond formations with hydroxylamines. These amide bond forming reactions proceed under aqueous conditions, without the need for coupling reagents or protecting groups. This product was introduced in collaboration with the Bode Research Group.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

804452-1G:
804452-BULK:
804452-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyler J Auvil et al.
Angewandte Chemie (International ed. in English), 52(43), 11317-11320 (2013-09-17)
It takes two: A unique organocatalyzed cascade for the unsymmetric double arylation of α-nitrodiazoesters is described. This organocascade features the strategic use of carbene-activating anilines in conjunction with a urea catalyst, thus allowing for the synthesis of pharmaceutically attractive α-diarylesters
David M Nickerson et al.
Chemical communications (Cambridge, England), 49(39), 4289-4291 (2012-12-01)
The strategic incorporation of internal Lewis acids onto urea scaffolds gives rise to a family of tunable hydrogen bond donor catalysts. The nature of the Lewis acid and associated ligands affects the urea polarization, acidity, and activity in reactions of
Sonia S So et al.
Organic letters, 14(2), 444-447 (2012-01-03)
Boronate ureas operate as catalysts for the activation of nitrocyclopropane carboxylates in nucleophilic ring-opening reactions. A variety of amines were found to open the urea-activated nitrocyclopropane carboxylates, generating highly useful nitro ester building blocks in good yields. Standard manipulations allow
Andrea M Hardman et al.
Organic & biomolecular chemistry, 11(35), 5793-5797 (2013-08-03)
Highly functionalized oxazinanes are efficiently prepared through urea-catalyzed formal [3 + 3] cycloaddition reactions of nitrones and nitrocyclopropane carboxylates. The reaction system is general with respect to both the nitrocyclopropane carboxylates and nitrones enabling the preparation of a large family
Sonia S So et al.
Organic letters, 13(4), 716-719 (2011-01-14)
Boronate ureas are introduced as a new class of noncovalent catalysts for conjugate addition reactions with enhanced activity. Through intramolecular coordination of the urea functionality to a strategically placed Lewis acid, rate enhancements up to 10 times that of more

Related Content

Bode Group creates N-mesityl-NHC catalysts for enantioselective annulations and SnAP reagents for one-pot aldehyde conversion into N-heterocycles.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service