Skip to Content
Merck
All Photos(1)

Documents

Safety Information

798495

Sigma-Aldrich

Titanium dioxide

22 nm, nanocrystalline colloidal paste for transparent film, >95% (anatase (XRD))

Synonym(s):

Nanocrystalline TiO2, TiO2 paste, Titania paste

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
UNSPSC Code:
12352302

description

Metals purity: ≤ 11 ppm Fe, Na and K (ICP)
Rheology at shear rate: 0.428 at 1000cm-1 (Pa.s)
Rheology at shear rate: 1850 at 0.01 cm-1 (Pa.s)

Assay

>95% (anatase (XRD))

form

paste (nanocrystalline colloid)

concentration

16 wt. %

particle size

22 nm
22-25 nm (BET)

pore size

21-22 nm average pore size (BET)
60-65 % porosity (BET)

pH

<1

conductivity

96.4 mS/cm

Looking for similar products? Visit Product Comparison Guide

Application

Titanium dioxide (TiO2) is used in the creation of photoelectrodes for the production of dye sensitized perovskite and other solid state solar cells. Our TiO2 pastes allow deposition of titania films with large surface to volume ratio. Select transparent TiO2 paste for applications that require a transparent sintered titania film and use opaque TiO2 paste for applications that do not require transparency. The opaque TiO2 paste contains larger scattering TiO2 particles.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

798495-VAR:
798495-25G:4548174005538
798495-BULK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service