Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

79422

Sigma-Aldrich

Phosphazene base P4-t-Oct solution

1.00 M±0.05 M in hexane

Synonym(s):

1-tert-Octyl-4,4,4-tris(dimethylamino)-2,2-bis[tris(dimethylamino)phosphoranylidenamino]-2λ5,4λ5-catenadi(phosphazene)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H71N13P4
CAS Number:
Molecular Weight:
689.82
Beilstein:
6641377
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.00 M±0.05 M in hexane

SMILES string

CN(C)P(=NP(=NC(C)(C)CC(C)(C)C)(N=P(N(C)C)(N(C)C)N(C)C)N=P(N(C)C)(N(C)C)N(C)C)(N(C)C)N(C)C

InChI

1S/C26H71N13P4/c1-25(2,3)24-26(4,5)27-40(28-41(31(6)7,32(8)9)33(10)11,29-42(34(12)13,35(14)15)36(16)17)30-43(37(18)19,38(20)21)39(22)23/h24H2,1-23H3

InChI key

HGZNCRLCTDLORC-UHFFFAOYSA-N

Other Notes

Extremely strong and hindered, neutral nitrogen base. It is about 1017 times more basic than DBU [pKa (DMSO) 29.43]

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Type 1 petroleums
Hazardous rank II
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

79422-1ML:4548173180021
79422-BULK:
79422-VAR:
79422-5ML:4548173180038


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Schwesinger et al.
Angewandte Chemie (International Edition in English), 99, 1212-1212 (1987)
R. Schwesinger et al
Angewandte Chemie (International Edition in English), 105, 1420-1420 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service